Anti-Retinoblastoma binding protein 1 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB25913-100EA 4490 DKK
ABCAAB25913-100
Anti-Retinoblastoma binding protein 1 Rabbit polyclonal antibody
Type: Primary
Antigen: Retinoblastoma binding protein 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR