Anti-CD8A Mouse monoclonal antibody [clone: RFT-8]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB25618-100EA 4300 DKK
ABCAAB25618-100
Anti-CD8A Mouse monoclonal antibody [clone: RFT-8]
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone: RFT-8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR