Anti-SELL Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: MEL-14]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB25569-50EA 2089.35 DKK
ABCAAB25569-50
Anti-SELL Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: MEL-14]
Antistoffer
Type: Primary
Antigen: SELL (selectin L)
Clonality: monoclonal
Clone: MEL-14
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR