Anti-CD3E Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: C363.29B]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB25531-50EA 4104.08 DKK
ABCAAB25531-50
Anti-CD3E Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: C363.29B]
Antistoffer
Type: Primary
Antigen: CD3E (CD3e molecule, epsilon (CD3-TCR complex))
Clonality: monoclonal
Clone: C363.29B
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Mouse