Anti-CD69 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: H1.2F3]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB25529-50EA 5630 DKK
ABCAAB25529-50
Anti-CD69 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: H1.2F3]
Antistoffer
Type: Primary
Antigen: CD69 (CD69 molecule)
Clonality: monoclonal
Clone: H1.2F3
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR