Anti-CD4 Mouse monoclonal antibody PE (Phycoerythrin)/Cy5.5® [clone: RFT-4g]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB25330-50EA 4650 DKK
ABCAAB25330-50
Anti-CD4 Mouse monoclonal antibody PE (Phycoerythrin)/Cy5.5® [clone: RFT-4g]
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone: RFT-4g
Conjugation: PE (Phycoerythrin)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR