Anti-CD3E Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: C363.29B (YCD3-1)]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB25278-50EA 4098.45 DKK
ABCAAB25278-50
Anti-CD3E Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: C363.29B (YCD3-1)]
Antistoffer
Type: Primary
Antigen: CD3E (CD3e molecule, epsilon (CD3-TCR complex))
Clonality: monoclonal
Clone: C363.29B (YCD3-1)
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR