Anti-LAMP1 Rat Monoclonal Antibody [clone: 1D4B]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB25245-100EA 6040 DKK
ABCAAB25245-100
Anti-LAMP1 Rat Monoclonal Antibody [clone: 1D4B]
Antistoffer
Type: Primary
Antigen: LAMP1 (lysosomal-associated membrane protein 1)
Clonality: monoclonal
Clone: 1D4B
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR