Anti-MHC Class 1 H2 Db Mouse Monoclonal Antibody [clone: 28-14-8]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB25228-100EA 5570 DKK
ABCAAB25228-100
Anti-MHC Class 1 H2 Db Mouse Monoclonal Antibody [clone: 28-14-8]
Antistoffer
Type: Primary
Antigen: MHC Class 1 H2 Db
Clonality: monoclonal
Clone: 28-14-8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR