Anti-CCL11 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB25086-100EA 5670 DKK
ABCAAB25086-100
Anti-CCL11 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: CCL11 (chemokine (C-C motif) ligand 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR