Anti-IL4 Mouse Monoclonal Antibody [clone: 5B5]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB25033-100EA 5650 DKK
ABCAAB25033-100
Anti-IL4 Mouse Monoclonal Antibody [clone: 5B5]
Antistoffer
Type: Primary
Antigen: IL4 (interleukin 4)
Clonality: monoclonal
Clone: 5B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR