Anti-NFKBIA Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB24784-50EA 4098.45 DKK
ABCAAB24784-50
Anti-NFKBIA Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NFKBIA (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human