Anti-PPP2R1A Rat monoclonal antibody [clone: 6G3]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24736-100EA 4300 DKK
ABCAAB24736-100
Anti-PPP2R1A Rat monoclonal antibody [clone: 6G3]
Type: Primary
Antigen: PPP2R1A (protein phosphatase 2, regulatory subunit A, alpha)
Clonality: monoclonal
Clone: 6G3
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR