Anti-Nuclear Pore Complex Proteins Mouse Monoclonal Antibody [clone: Mab414]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24609-100EA 4514.49 DKK
ABCAAB24609-100
Anti-Nuclear Pore Complex Proteins Mouse Monoclonal Antibody [clone: Mab414]
Antistoffer
Type: Primary
Antigen: Nuclear Pore Complex Proteins
Clonality: monoclonal
Clone: Mab414
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR