Anti-GOLGB1 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24586-100EA 4610 DKK
ABCAAB24586-100
Anti-GOLGB1 Rabbit polyclonal antibody
Type: Primary
Antigen: GOLGB1 (golgin B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR