Anti-200kDa + 160kDa Neurofilament Mouse monoclonal antibody [clone: SMI-310]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24570-250EA 4300 DKK
ABCAAB24570-250
Anti-200kDa + 160kDa Neurofilament Mouse monoclonal antibody [clone: SMI-310]
Type: Primary
Antigen: 200kDa + 160kDa Neurofilament
Clonality: monoclonal
Clone: SMI-310
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR