Anti-KRT18 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24561-100EA 3954.84 DKK
ABCAAB24561-100
Anti-KRT18 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR