Anti-CD16 + CD32 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: FCR 4G8]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24537-50EA 2740 DKK
ABCAAB24537-50
Anti-CD16 + CD32 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: FCR 4G8]
Antistoffer
Type: Primary
Antigen: CD16 + CD32
Clonality: monoclonal
Clone: FCR 4G8
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR