Anti-FADD Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24533-100EA 5810 DKK
ABCAAB24533-100
Anti-FADD Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: FADD (Fas (TNFRSF6)-associated via death domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR