Anti-CTSV Mouse Monoclonal Antibody [clone: BV55-1]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB24508-100EA 0 DKK
ABCAAB24508-100
Anti-CTSV Mouse Monoclonal Antibody [clone: BV55-1]
Antistoffer
Type: Primary
Antigen: CTSV (cathepsin V)
Clonality: monoclonal
Clone: BV55-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human