Anti-CD44 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24504-100EA 4061.19 DKK
ABCAAB24504-100
Anti-CD44 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR