Anti-CD5 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: OX-19]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24500-50EA 5630 DKK
ABCAAB24500-50
Anti-CD5 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: OX-19]
Antistoffer
Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: monoclonal
Clone: OX-19
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR