Anti-Serum Albumin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 1A9]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24438-200EA 5030 DKK
ABCAAB24438-200
Anti-Serum Albumin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 1A9]
Antistoffer
Type: Primary
Antigen: Serum Albumin
Clonality: Monoclonal
Clone: 1A9
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR