Anti-LRP6 Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24386-100EA 5670 DKK
ABCAAB24386-100
Anti-LRP6 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LRP6 (low density lipoprotein receptor-related protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR