Anti-HLA G Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-G/9]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24384-100EA 5760 DKK
ABCAAB24384-100
Anti-HLA G Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-G/9]
Antistoffer
Type: Primary
Antigen: HLA G
Clonality: monoclonal
Clone: MEM-G/9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR