Anti-Retinoid X Receptor gamma Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24364-100EA 5370 DKK
ABCAAB24364-100
Anti-Retinoid X Receptor gamma Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Retinoid X Receptor gamma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR