Anti-NRCAM Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24344-200EA 4098.45 DKK
ABCAAB24344-200
Anti-NRCAM Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NRCAM (neuronal cell adhesion molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR