Anti-SUC1 Mouse Monoclonal Antibody [clone: AA10.2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24294-50EA 5370 DKK
ABCAAB24294-50
Anti-SUC1 Mouse Monoclonal Antibody [clone: AA10.2]
Antistoffer
Type: Primary
Antigen: SUC1
Clonality: monoclonal
Clone: AA10.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR