Anti-IDUA Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24227-100EA 5430 DKK
ABCAAB24227-100
Anti-IDUA Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: IDUA (iduronidase, alpha-L-)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR