Anti-Blood Group N antigen Mouse Monoclonal Antibody [clone: DRF-8]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24217-100EA 5370 DKK
ABCAAB24217-100
Anti-Blood Group N antigen Mouse Monoclonal Antibody [clone: DRF-8]
Antistoffer
Type: Primary
Antigen: Blood Group N antigen
Clonality: monoclonal
Clone: DRF-8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR