Anti-Substance P Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24126-125EA 4060 DKK
ABCAAB24126-125
Anti-Substance P Rabbit polyclonal antibody
Type: Primary
Antigen: Substance P
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR