Anti-COL1/2/3/4/5 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24117-500EA 5420 DKK
ABCAAB24117-500
Anti-COL1/2/3/4/5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: COL1/2/3/4/5 (collagen, type I/II/III/IV/V)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR