Anti-IFNA2 Mouse Monoclonal Antibody [clone: N27]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB24099-100EA 5330 DKK
ABCAAB24099-100
Anti-IFNA2 Mouse Monoclonal Antibody [clone: N27]
Antistoffer
Type: Primary
Antigen: IFNA2 (interferon, alpha 2)
Clonality: monoclonal
Clone: N27
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR