Anti-Kanamycin Receptor Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB23971-100EA 3800.38 DKK
ABCAAB23971-100
Anti-Kanamycin Receptor Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Kanamycin Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu