Anti-HIST2H2AC Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB23957-100EA 2710 DKK
ABCAAB23957-100
Anti-HIST2H2AC Rabbit polyclonal antibody
Type: Primary
Antigen: HIST2H2AC (histone cluster 2, H2ac)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR