Anti-Maltose Binding Protein Mouse Monoclonal Antibody [clone: R29]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB23903-125EA 5330 DKK
ABCAAB23903-125
Anti-Maltose Binding Protein Mouse Monoclonal Antibody [clone: R29]
Antistoffer
Type: Primary
Antigen: Maltose Binding Protein
Clonality: monoclonal
Clone: R29
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR