Anti-ITGB1 Armenian Hamster monoclonal antibody [clone: HM beta 1.1]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB23833-100EA 4060 DKK
ABCAAB23833-100
Anti-ITGB1 Armenian Hamster monoclonal antibody [clone: HM beta 1.1]
Type: Primary
Antigen: ITGB1 (integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12))
Clonality: monoclonal
Clone: HM beta 1.1
Conjugation: unconjugated
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR