Anti-FOXA1 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB23738-100EA 5930 DKK
ABCAAB23738-100
Anti-FOXA1 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: FOXA1 (forkhead box A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR