Anti-CD99 Mouse Monoclonal Antibody [clone: DN16]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB23617-100EA 4098.45 DKK
ABCAAB23617-100
Anti-CD99 Mouse Monoclonal Antibody [clone: DN16]
Antistoffer
Type: Primary
Antigen: CD99 (CD99 molecule)
Clonality: monoclonal
Clone: DN16
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR