Anti-CD82 Mouse monoclonal antibody [clone: B-L2]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB23525-100EA 4060 DKK
ABCAAB23525-100
Anti-CD82 Mouse monoclonal antibody [clone: B-L2]
Type: Primary
Antigen: CD82 (CD82 molecule)
Clonality: monoclonal
Clone: B-L2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR