Anti-HSV1 + HSV2 Mouse monoclonal antibody [clone: 0119]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB23494-500EA 4140 DKK
ABCAAB23494-500
Anti-HSV1 + HSV2 Mouse monoclonal antibody [clone: 0119]
Type: Primary
Antigen: HSV1 + HSV2
Clonality: monoclonal
Clone: 119
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR