Anti-Phencyclidine Mouse monoclonal antibody [clone: 6E7]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB23358-200EA 4250 DKK
ABCAAB23358-200
Anti-Phencyclidine Mouse monoclonal antibody [clone: 6E7]
Type: Primary
Antigen: Phencyclidine
Clonality: monoclonal
Clone: 60000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR