Anti-Proteasome 20S alpha 7 Mouse monoclonal antibody [clone: MCP34]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB22669-50EA 4140 DKK
ABCAAB22669-50
Anti-Proteasome 20S alpha 7 Mouse monoclonal antibody [clone: MCP34]
Type: Primary
Antigen: Proteasome 20S alpha 7
Clonality: monoclonal
Clone: MCP34
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR