Anti-hCG alpha 3 Mouse monoclonal antibody [clone: INN-hCG-5]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB22488-100EA 4140 DKK
ABCAAB22488-100
Anti-hCG alpha 3 Mouse monoclonal antibody [clone: INN-hCG-5]
Type: Primary
Antigen: hCG alpha 3
Clonality: monoclonal
Clone: INN-hCG-5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR