Anti-CD19 Rat Monoclonal Antibody (Biotin) [clone: 6D5]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB22477-100EA 5330 DKK
ABCAAB22477-100
Anti-CD19 Rat Monoclonal Antibody (Biotin) [clone: 6D5]
Antistoffer
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone: 6D5
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR