Anti-FOXP3 Rabbit polyclonal antibody HRP (Horseradish Peroxidase)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB22273-100EA 4060 DKK
ABCAAB22273-100
Anti-FOXP3 Rabbit polyclonal antibody HRP (Horseradish Peroxidase)
Type: Primary
Antigen: FOXP3 (forkhead box P3)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR