Anti-MYOCD Mouse polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB22073-100EA 4690 DKK
ABCAAB22073-100
Anti-MYOCD Mouse polyclonal antibody
Type: Primary
Antigen: MYOCD (myocardin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR