Anti-GAD1 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB22050-100EA 4690 DKK
ABCAAB22050-100
Anti-GAD1 Rabbit polyclonal antibody
Type: Primary
Antigen: GAD1 (glutamate decarboxylase 1 (brain, 67kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR