Anti-IVa2 Mouse polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB21939-100EA 4140 DKK
ABCAAB21939-100
Anti-IVa2 Mouse polyclonal antibody
Type: Primary
Antigen: IVa2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR