Anti-ICAM3 Mouse monoclonal antibody [clone: MEM-04]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB21900-100EA 4250 DKK
ABCAAB21900-100
Anti-ICAM3 Mouse monoclonal antibody [clone: MEM-04]
Type: Primary
Antigen: ICAM3 (intercellular adhesion molecule 3)
Clonality: monoclonal
Clone: MEM-04
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR